missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzylamine, 97%, Thermo Scientific™

Catalog Number p-3629701
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
946409-19-4
C10H11N3O
189.218
MFCD11109325
SBNUQIHHPTZLSV-UHFFFAOYSA-N
4-5-methyl-1,3,4-oxadiazol-2-yl benzylamine, 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, 1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methylamine, 4-5-methyl-1,3,4-oxadiazol-2-yl-benzenemethanamine
33589501
[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine
CC1=NN=C(O1)C2=CC=C(C=C2)CN
This item is not returnable. View return policy
946409-19-4
C10H11N3O
189.218
MFCD11109325
SBNUQIHHPTZLSV-UHFFFAOYSA-N
4-5-methyl-1,3,4-oxadiazol-2-yl benzylamine, 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, 1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methylamine, 4-5-methyl-1,3,4-oxadiazol-2-yl-benzenemethanamine
33589501
[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine
CC1=NN=C(O1)C2=CC=C(C=C2)CN
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.22
Percent Purity 97%
Chemical Name or Material 4-(5-Methyl-1, 3, 4-oxadiazol-2-yl)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'