missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10612485
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10612485 5 g 5g
10361623 250 mg 250mg
10443223 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10612485 Supplier Thermo Scientific Maybridge Supplier No. CC34804DE
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-60-0
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD07772818
InChI Key BJICSBRPQDVEHP-UHFFFAOYSA-N
Synonym 4-5-methyl-1,2,4-oxadiazol-3-yl benzaldehyde, benzaldehyde,4-5-methyl-1,2,4-oxadiazol-3-yl
PubChem CID 7164589
IUPAC Name 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzaldehyde
SMILES CC1=NC(=NO1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 4-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.