missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Iodo-2-pyridyl)morpholine, 96%, Thermo Scientific™

Catalog Number 11494610
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
470463-42-4
C9H11IN2O
290.104
MFCD04039866
RBMHFKCAFPUVRT-UHFFFAOYSA-N
4-5-iodo-2-pyridinyl morpholine, 4-5-iodopyridin-2-yl morpholine, 4-5-iodo-pyridin-2-yl-morpholine, 4-5-iodo-2-pyridyl morpholine, 4-5-iodopyridine-2-yl morpholine, 4-5-iodo-pyridin-2-yl morpholine, 5-iodo-2-morpholin-4-yl pyridine, morpholine, 4-5-iodo-2-pyridinyl
2795362
4-(5-iodopyridin-2-yl)morpholine
C1COCCN1C2=NC=C(C=C2)I
This item is not returnable. View return policy
470463-42-4
C9H11IN2O
290.104
MFCD04039866
RBMHFKCAFPUVRT-UHFFFAOYSA-N
4-5-iodo-2-pyridinyl morpholine, 4-5-iodopyridin-2-yl morpholine, 4-5-iodo-pyridin-2-yl-morpholine, 4-5-iodo-2-pyridyl morpholine, 4-5-iodopyridine-2-yl morpholine, 4-5-iodo-pyridin-2-yl morpholine, 5-iodo-2-morpholin-4-yl pyridine, morpholine, 4-5-iodo-2-pyridinyl
2795362
4-(5-iodopyridin-2-yl)morpholine
C1COCCN1C2=NC=C(C=C2)I

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 131°C to 135°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 290.11
Percent Purity 96%
Chemical Name or Material 4-(5-Iodo-2-pyridyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'