missing translation for 'onlineSavingsMsg'
Learn More

4,5-Disulfo Rhodamine-123 Dicarboxylic Acid Lithium Salt(Mixture of isomers, Technical Grade), TRC

Product Code. 30383563
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383563 0.5 mg 0.50mg
30387142 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383563 Supplier Toronto Research Chemicals Supplier No. TRCD6788200.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 247144-99-6
Molecular Formula C22H16Li2N2O11S2
Molecular Weight (g/mol) 562.37
Synonym 3,6-Diamino-9-[2,4(or 2,5)-dicarboxyphenyl]-4,5-disulfoxanthylium Inner Salt Lithium Salt
SMILES [Li+].[Li+].[Li+].[*]C(=O)[O-].[*]c1cc(C(=O)[O-])c(cc1[*])c2c3ccc(N)c(c3[o+]c4c(c(N)ccc24)S(=O)(=O)[O-])S(=O)(=O)[O-].[*H]

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C20H14N2O9S2.C2H4O2.2Li/c21-13-7-5-11-15(9-3-1-2-4-10(9)20(23)24)12-6-8-14(22)19(33(28,29)30)17(12)31-16(11)18(13)32(25,26)27;1-2(3)4;;/h1-8,21H,22H2,(H,23,24)(H,25,26,27)(H,28,29,30);1H3,(H,3,4);;/q;;2*+1/p-2
Formula Weight 552.0284
Chemical Name or Material 4,5-Disulfo Rhodamine-123 Dicarboxylic Acid Lithium Salt(Mixture of isomers, Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.