missing translation for 'onlineSavingsMsg'
Learn More

4,5-Diphenylimidazole, 98+%

Product Code. 11482767
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
Product Code. Quantity unitSize
11482767 5 g 5g
11402777 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11482767

Brand: Thermo Scientific Alfa Aesar A14939.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 668-94-0 | C15H12N2 | 220.28 g/mol

4,5-Diphenylimidazole is used as a precursor for azo dyes such as 2-(quinolylazo)-4,5-diphenylimidazole . It is also used to determine metal ions by spectrophotometrically. It acts as a chromogenic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Diphenylimidazole is used as a precursor for azo dyes such as 2-(quinolylazo)-4,5-diphenylimidazole . It is also used to determine metal ions by spectrophotometrically. It acts as a chromogenic reagent.

Solubility
Soluble in methanol.

Notes
Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 668-94-0
Molecular Formula C15H12N2
Molecular Weight (g/mol) 220.28
MDL Number MFCD00005198
InChI Key CPHGOBGXZQKCKI-UHFFFAOYSA-N
Synonym 4,5-diphenylimidazole, diphenylimidazole, 1h-imidazole, 4,5-diphenyl, imidazole, 4,5-diphenyl, dsstox_cid_25700, dsstox_rid_81072, dsstox_gsid_45700, 4,5-diphenyl-1h-imidazol, imidazole,5-diphenyl, 4,5-diphenylglyoxaline
PubChem CID 69588
IUPAC Name 4,5-diphenyl-1H-imidazole
SMILES N1C=NC(=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 230°C to 234°C
Quantity 5 g
Beilstein 157587
Solubility Information Soluble in methanol.
Formula Weight 220.28
Percent Purity ≥98%
Chemical Name or Material 4,5-Diphenylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.