missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethylthiophene-2-carboxaldehyde, 97%

Product Code. 15483757
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483757 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15483757 Supplier Thermo Scientific Alfa Aesar Supplier No. H27712.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5928-48-3 | C7H8OS | 140.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5928-48-3
Density 1.179
Boiling Point 82°C to 85°C
Flash Point >110°C (230°F)
Molecular Formula C7H8OS
Refractive Index 1.579
MDL Number MFCD02093897
Quantity 1 g
Sensitivity Air Sensitive
Synonym 4,5-dimethylthiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 4,5-dimethyl, acmc-20amru, pubchem20559, 2,3-dimethyl-5-formylthiophene, 2-thiophenecarboxaldehyde,4,5-dimethyl, 4,5-dimethyl-2-thiophene carboxaldehyde
InChI Key QSBBXKVGLJSGAJ-UHFFFAOYSA-N
SMILES CC1=C(SC(=C1)C=O)C
IUPAC Name 4,5-dimethylthiophene-2-carbaldehyde
Molecular Weight (g/mol) 140.2
PubChem CID 2733926
Formula Weight 140.2
Percent Purity 97%
Chemical Name or Material 4,5-Dimethylthiophene-2-carboxaldehyde
Show More Show Less

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.