missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-2-furoic acid, 97%, Thermo Scientific™

Product Code. 10314612
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10314612 1 g Amber Glass Bottle 1g
10304612 250 mg Amber Glass Bottle 250mg
10326834 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10314612 Supplier Thermo Scientific Maybridge Supplier No. BTB08890DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89639-83-8
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00085031
InChI Key BNVXYXRSJIWWDX-UHFFFAOYSA-N
PubChem CID 2774924
IUPAC Name 4,5-dimethylfuran-2-carboxylic acid
SMILES CC1=C(OC(=C1)C(=O)O)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4,5-dimethyl-2-furoic acid, 2-furancarboxylic acid,4,5-dimethyl, 2-furancarboxylic acid, 4,5-dimethyl, 3-methylbenzofurane, acmc-20lon1, 4,5-dimethylfurancarboxylic acid, 4,5-dimethyl-2-furancarboxylic acid, 4,5-dimethyl-furan-2-carboxylic acid
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 4, 5-Dimethyl-2-furoic acid
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.