missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl chloroformate, 97%

Product Code. p-7046395
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15478999

Brand: Thermo Scientific Alfa Aesar L14167.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42855-00-5 | C10H10ClNO6 | 275.641 g/mol

4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, acetone. Reacts with water.

Notes
Moisture and light sensitive. Store away from oxidizing agents, light, water/ moisture, heat, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42855-00-5
Molecular Formula C10H10ClNO6
Molecular Weight (g/mol) 275.641
MDL Number MFCD00143507
InChI Key RWWPKIOWBQFXEE-UHFFFAOYSA-N
Synonym 4,5-dimethoxy-2-nitrobenzyl carbonochloridate, nvoc-cl, 4,5-dimethoxy-2-nitrobenzyl chloroformate, 6-nitroveratryl chloroformate, nitroveratryl oxy chlorocarbamate, 6-nitroveratryloxycarbonyl chloride, carbonochloridic acid, 4,5-dimethoxy-2-nitrophenyl methyl ester, 4,5-dimethoxy-2-nitrophenyl methyl chloroformate, 4,5-dimethoxy-2-nitrophenyl methyl carbonochloridate, nvoc chloride
PubChem CID 3084878
IUPAC Name (4,5-dimethoxy-2-nitrophenyl)methyl carbonochloridate
SMILES COC1=C(C=C(C(=C1)COC(=O)Cl)[N+](=O)[O-])OC

Specifications

Melting Point ∼120°C
Quantity 250 mg
UN Number UN3261
Beilstein 2389168
Formula Weight 275.64
Percent Purity 97%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzyl chloroformate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.