missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic Acid, TRC

Product Code. 30380836
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380836 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380836 Supplier Toronto Research Chemicals Supplier No. TRCD44912510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid
CAS 72909-34-3
Molecular Formula C14 H6 N2 O8
Quantity 10 mg
Synonym 1H-Pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, 4,5-dihydro-4,5-dioxo-, 4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, 2,7,9-Tricarboxy-4,5-dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline, Coenzyme PQQ, Methoxatin, Methoxatine, PQQ, Pyrroloquinoline quinone
InChI Formula InChI=1S/C14H6N2O8/c17-10-4-2-6(14(23)24)15-8(4)7-3(12(19)20)1-5(13(21)22)16-9(7)11(10)18/h1-2,15H,(H,19,20)(H,21,22)(H,23,24)
SMILES OC(=O)c1cc(C(=O)O)c2c(n1)C(=O)C(=O)c3cc([nH]c23)C(=O)O
IUPAC Name 4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid
Molecular Weight (g/mol) 330.21
Formula Weight 330.0124
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.