missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-nitrobenzoic acid, 99%, Thermo Scientific™

Product Code. 11985011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11985011 5 g 5g
11975011 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985011 Supplier Thermo Scientific Acros Supplier No. 442670050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 20372-63-8
Molecular Formula C7H3F2NO4
Molecular Weight (g/mol) 203.1
InChI Key HGGRAOYTQNFGGN-UHFFFAOYSA-N
Synonym 4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 3,4-difluoro-6-nitrobenzoic acid, 2-nitro-4,5-difluorobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, benzoic acid, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-two fluoro-2-nitrobenzoic acid, pubchem4947, acmc-1cegm
PubChem CID 4219630
SMILES C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 166.0°C to 167.0°C
Assay Percent Range 98.5% min. (HPLC)
Quantity 5 g
Formula Weight 203.1
Percent Purity 99%
Chemical Name or Material 4, 5-Difluoro-2-nitrobenzoic acid
compliance-icons
Product Identifier
  • 4,5-Difluoro-2-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.