missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-nitroaniline, 98%

Product Code. 15492188
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492188 1 g 1g
15412198 250 mg 250mg
15402198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492188 Supplier Thermo Scientific Alfa Aesar Supplier No. H55830.03
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78056-39-0 | C6H4F2N2O2 | 174.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78056-39-0
Molecular Formula C6H4F2N2O2
Molecular Weight (g/mol) 174.11
MDL Number MFCD00010876
InChI Key WDMCABATCGQAKK-UHFFFAOYSA-N
Synonym 3,4-difluoro-6-nitroaniline, benzenamine, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitro-aniline, 4,5-difluoro-2-nitrophenylamine, 2-nitro-4,5-difluoroaniline, 2-amino-4,5-difluoronitrobenzene, 4,5-difluoro-2-nitro-phenylamine, 4,5-difluoro-2-nitrobenzenamine, pubchem2121, acmc-209pdb
PubChem CID 157151
IUPAC Name 4,5-difluoro-2-nitroaniline
SMILES NC1=CC(F)=C(F)C=C1[N+]([O-])=O

Specifications

Melting Point 107°C to 108°C
Quantity 1 g
Beilstein 2723242
Formula Weight 174.11
Percent Purity 98%
Chemical Name or Material 4,5-Difluoro-2-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.