missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-methoxybenzonitrile, 99%, Thermo Scientific™

Product Code. 15403427
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 425702-28-9
Molecular Formula C8H5F2NO
Molecular Weight (g/mol) 169.131
MDL Number MFCD06660346
InChI Key HACKETIBYOIPCJ-UHFFFAOYSA-N
Synonym 2-methoxy-4,5-difluorobenzonitrile, 2-cyano-4,5-difluoroanisole, benzonitrile, 4,5-difluoro-2-methoxy, 4,5-difluoro-2-methoxy-benzonitrile, 4,5-difluoro-2-methoxybenzenecarbonitrile, pubchem4767, acmc-1ao6r, ksc235i9n, 4,5-difluoro-2-methoxybenzonitrile
PubChem CID 12040834
IUPAC Name 4,5-difluoro-2-methoxybenzonitrile
SMILES COC1=CC(=C(C=C1C#N)F)F

Specifications

Melting Point 98°C to 100°C
Quantity 250 mg
UN Number UN3439
Formula Weight 169.13
Percent Purity 99%
Chemical Name or Material 4,5-Difluoro-2-methoxybenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'