missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-hydroxybenzonitrile, 99%, Thermo Scientific Chemicals

Product Code. p-7028749
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15493417

Brand: Thermo Scientific Alfa Aesar H26155.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4,5-Difluoro-2-hydroxybenzonitrile crystals have been used as a practical means for monitoring, imaging, and other purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Difluoro-2-hydroxybenzonitrile crystals have been used as a practical means for monitoring, imaging, and other purposes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186590-36-3
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD08275287
InChI Key ROLMZTIHUMKEAI-UHFFFAOYSA-N
Synonym 4,5-difluoro-2-hydroxy-benzonitrile, benzonitrile, 4,5-difluoro-2-hydroxy, acmc-1c7c6, 2-cyano-4,5-difluorophenol, 4,5-difluoro-2-hydroxybenzenecarbonitrile, benzonitrile, 4,5-difluoro-2-hydroxy-9ci
PubChem CID 23148985
IUPAC Name 4,5-difluoro-2-hydroxybenzonitrile
SMILES OC1=CC(F)=C(F)C=C1C#N

Specifications

Melting Point 161°C to 163°C
Quantity 250 mg
UN Number UN3439
Formula Weight 155.1
Percent Purity 99%
Chemical Name or Material 4,5-Difluoro-2-hydroxybenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.