missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-hydroxybenzonitrile, 99%, Thermo Scientific Chemicals

Product Code. 11326529
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326529 1 g 1g
15493417 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326529 Supplier Thermo Scientific Alfa Aesar Supplier No. H26155.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 186590-36-3 | Molecular Formula: C7H3F2NO | Molecular Weight: 155.10 g/mol

4,5-Difluoro-2-hydroxybenzonitrile crystals have been used as a practical means for monitoring, imaging, and other purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Difluoro-2-hydroxybenzonitrile crystals have been used as a practical means for monitoring, imaging, and other purposes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186590-36-3
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD08275287
InChI Key ROLMZTIHUMKEAI-UHFFFAOYSA-N
Synonym 4,5-difluoro-2-hydroxy-benzonitrile, benzonitrile, 4,5-difluoro-2-hydroxy, acmc-1c7c6, 2-cyano-4,5-difluorophenol, 4,5-difluoro-2-hydroxybenzenecarbonitrile, benzonitrile, 4,5-difluoro-2-hydroxy-9ci
PubChem CID 23148985
IUPAC Name 4,5-difluoro-2-hydroxybenzonitrile
SMILES OC1=CC(F)=C(F)C=C1C#N

Specifications

Melting Point 161°C to 163°C
Quantity 1 g
UN Number UN3439
Formula Weight 155.1
Percent Purity 99%
Chemical Name or Material 4,5-Difluoro-2-hydroxybenzonitrile

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.