missing translation for 'onlineSavingsMsg'
Learn More

4,5-Didehydro Brimonidine, TRC

Product Code. 30133693
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133693 1 g 1g
30133752 250 mg 250mg
18050350 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133693 Supplier Toronto Research Chemicals Supplier No. TRCB6871801G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151110-15-5
Molecular Formula C11 H8 Br N5
Molecular Weight (g/mol) 290.1187
Synonym 5-Bromo-N-1H-imidazol-2-yl-6-quinoxalinamine, 5-Bromo-N-(1H-imidazol-2-yl)quinoxalin-6-amine, Brimonidine EP Impurity F
IUPAC Name 5-bromo-N-(1H-imidazol-2-yl)quinoxalin-6-amine
SMILES Brc1c(Nc2ncc[nH]2)ccc3nccnc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H8BrN5/c12-9-7(17-11-15-5-6-16-11)1-2-8-10(9)14-4-3-13-8/h1-6H,(H2,15,16,17)
Formula Weight 288.9963
Percent Purity >95%
Chemical Name or Material 4,5-Didehydro Brimonidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.