missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dicyanoimidazole, TRC

Product Code. 30384721
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384721 25 g 25g
30381616 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384721 Supplier Toronto Research Chemicals Supplier No. TRCD43775525G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,5-Dicyanoimidazole
CAS 1122-28-7
Molecular Formula C5H2N4
Quantity 25 g
Synonym 1H-Imidazole-4,5-dicarbonitrile, Imidazole-4,5-dicarbonitrile, 2H-Imidazole-4,5-dicarbonitrile, MOA 7, NSC 113954
InChI Formula InChI=1S/C5H2N4/c6-1-4-5(2-7)9-3-8-4/h3H,(H,8,9)
SMILES N#Cc1nc[nH]c1C#N
Molecular Weight (g/mol) 118.1
Formula Weight 118.0279
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.