missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichlorophthalic Anhydride, TRC

Product Code. 30371089
Change view
Click to view available options
Quantity:
1 g
5 g
Product Code. Quantity
30371089 5 g
30370087 1 g
2 options
This item is not returnable. View return policy

Product Code. 30371089

Brand: Toronto Research Chemicals TRCD4739635G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942-06-3
Formula Weight 215.9381
InChI Formula InChI=1S/C8H2Cl2O3/c9-5-1-3-4(2-6(5)10)8(12)13-7(3)11/h1-2H
IUPAC Name 5,6-dichloro-2-benzofuran-1,3-dione
Molecular Formula C8H2Cl2O3
Molecular Weight (g/mol) 217.01
SMILES Clc1cc2C(=O)OC(=O)c2cc1Cl
Synonym 5,6-Dichloroisobenzofuran-1,3-dione
Chemical Name or Material 4,5-Dichlorophthalic Anhydride

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.