missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichlorocatechol, TRC

Product Code. 30385169
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385169 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385169 Supplier Toronto Research Chemicals Supplier No. TRCD43400010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,5-Dichlorocatechol
CAS 3428-24-8
Molecular Formula C6 H4 Cl2 O2
Quantity 10 g
Synonym Pyrocatechol, 4,5-dichloro- (7CI,8CI), 4,5-Dichloro-1,2-benzenediol, 4,5-Dichloro-1,2-dihydroxybenzene, 4,5-Dichlorocatechol, 4,5-Dichloropyrocatechol
InChI Formula InChI=1S/C6H4Cl2O2/c7-3-1-5(9)6(10)2-4(3)8/h1-2,9-10H
SMILES Oc1cc(Cl)c(Cl)cc1O
IUPAC Name 4,5-dichlorobenzene-1,2-diol
Molecular Weight (g/mol) 179
Formula Weight 177.9588
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.