missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-o-phenylenediamine, 98%

Product Code. 11493328
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493328 5 g 5g
11403338 25 g 25g
11413338 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493328 Supplier Thermo Scientific Alfa Aesar Supplier No. A17738.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5348-42-5 | C6H6Cl2N2 | 177.03 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol and Dimethyl sulfoxide. Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5348-42-5
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.03
MDL Number MFCD00007723
InChI Key IWFHBRFJOHTIPU-UHFFFAOYSA-N
Synonym 4,5-dichloro-o-phenylenediamine, 4,5-dichloro-1,2-benzenediamine, 1,2-diamino-4,5-dichlorobenzene, 4,5-dichloro-1,2-phenylenediamine, 1,2-benzenediamine, 4,5-dichloro, 4,5-dichloro-o-phenylendiamine, 1,2-dichloro-4,5-diaminobenzene, 2-amino-4,5-dichloroaniline, 4,5-dichloro-ortho-phenylenediamine, 4,5-dichlorophenylene-1,2-diamine
PubChem CID 79297
IUPAC Name 4,5-dichlorobenzene-1,2-diamine
SMILES NC1=CC(Cl)=C(Cl)C=C1N

Specifications

Melting Point ∼162°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 1072811
Solubility Information Soluble in methanol and Dimethyl sulfoxide. Insoluble in water.
Formula Weight 177.03
Percent Purity 98%
Chemical Name or Material 4,5-Dichloro-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.