missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dichloro-2-octyl-3(2H)-isothiazolone, TRC
Specifications
Specifications
| Chemical Name or Material | 4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one |
| CAS | 64359-81-5 |
| Molecular Formula | C11 H17 Cl2 N O S |
| Quantity | 500 mg |
| Synonym | 4,5-Dichloro-2-octyl-3(2H)-isothiazolone, 2-(n-Octyl)-dichloro-3-isothiazolone, 4,5-Dichloro-2-N-octyl-3(2H)-isothiazolone, 4,5-Dichloro-2-n-octyl-3-isothiazolone, 4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one, 4,5-Dichloro-2-n-octyl-4-isothiazoline-3-one, 4,5-Dichloro-2-n-octylisothiazolin-3-one, 4,5-Dichloro-2-octyl-3-isothiazolone, 4,5-Dichloro-2-octyl-4-isothiazolin-3-one, 4,5-Dichloro-2-octyl-isothiazolone, 4,5-Dichloro-2-octylisothiazolin-3-one, 4,5-Dichloro-n-octylisothiazolin-3-one, Bioban 200, C 9211, DC 30, DC 30 (antibacterial), DCOIT, DCOIT 30, DCOIT 97, Dichloro-N-octylisothiazolin-3-one, Dichlorooctylisothiazolinone, Ecoplast T 20, Kathon 287, Kathon 287PXE, Kathon 287T, Kathon 287tech, Kathon 5287, Kathon 910SB, Kathon 930, Kathon RH 287, Klarix 4000, Nalco 2894, RH 287, Rocima 200, Rozone 2000, Sea-Nine, Sea-Nine 211, Sea-Nine 211N, Sea-Nine 221, Sea-Nine CR 2, SeaNine CR Marine Anti-Fouling Agent, Vinyzene DCOIT |
| InChI Formula | InChI=1S/C11H17Cl2NOS/c1-2-3-4-5-6-7-8-14-11(15)9(12)10(13)16-14/h2-8H2,1H3 |
| SMILES | CCCCCCCCN1SC(=C(Cl)C1=O)Cl |
| IUPAC Name | 4,5-dichloro-2-octyl-1,2-thiazol-3-one |
| Molecular Weight (g/mol) | 282.23 |
| Formula Weight | 281.0408 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?