missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-octyl-3(2H)-isothiazolone, TRC

Product Code. 30382088
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382088 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382088 Supplier Toronto Research Chemicals Supplier No. TRCD435923100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one
CAS 64359-81-5
Molecular Formula C11 H17 Cl2 N O S
Quantity 100 mg
Synonym 4,5-Dichloro-2-octyl-3(2H)-isothiazolone, 2-(n-Octyl)-dichloro-3-isothiazolone, 4,5-Dichloro-2-N-octyl-3(2H)-isothiazolone, 4,5-Dichloro-2-n-octyl-3-isothiazolone, 4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one, 4,5-Dichloro-2-n-octyl-4-isothiazoline-3-one, 4,5-Dichloro-2-n-octylisothiazolin-3-one, 4,5-Dichloro-2-octyl-3-isothiazolone, 4,5-Dichloro-2-octyl-4-isothiazolin-3-one, 4,5-Dichloro-2-octyl-isothiazolone, 4,5-Dichloro-2-octylisothiazolin-3-one, 4,5-Dichloro-n-octylisothiazolin-3-one, Bioban 200, C 9211, DC 30, DC 30 (antibacterial), DCOIT, DCOIT 30, DCOIT 97, Dichloro-N-octylisothiazolin-3-one, Dichlorooctylisothiazolinone, Ecoplast T 20, Kathon 287, Kathon 287PXE, Kathon 287T, Kathon 287tech, Kathon 5287, Kathon 910SB, Kathon 930, Kathon RH 287, Klarix 4000, Nalco 2894, RH 287, Rocima 200, Rozone 2000, Sea-Nine, Sea-Nine 211, Sea-Nine 211N, Sea-Nine 221, Sea-Nine CR 2, SeaNine CR Marine Anti-Fouling Agent, Vinyzene DCOIT
InChI Formula InChI=1S/C11H17Cl2NOS/c1-2-3-4-5-6-7-8-14-11(15)9(12)10(13)16-14/h2-8H2,1H3
SMILES CCCCCCCCN1SC(=C(Cl)C1=O)Cl
IUPAC Name 4,5-dichloro-2-octyl-1,2-thiazol-3-one
Molecular Weight (g/mol) 282.23
Formula Weight 281.0408
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.