missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-nitroaniline, TRC

Product Code. 30369417
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369417 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369417 Supplier Toronto Research Chemicals Supplier No. TRCD4351702.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 4,5-Dichloro-2-nitroaniline
CAS 6641-64-1
Molecular Formula C6H4Cl2N2O2
Quantity 2.5 g
InChI Formula InChI=1S/C6H4Cl2N2O2/c7-3-1-5(9)6(10(11)12)2-4(3)8/h1-2H,9H2
SMILES Nc1cc(Cl)c(Cl)cc1[N+](=O)[O-]
Molecular Weight (g/mol) 207.01
Formula Weight 205.965
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.