missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 15433167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433167 5 g 5g
15423167 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433167 Supplier Thermo Scientific Alfa Aesar Supplier No. A17908.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6641-64-1 | Molecular Formula: C6H4Cl2N2O2 | Molecular Weight: 207.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6641-64-1
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.01
MDL Number MFCD00007770
InChI Key FSGTULQLEVAYRS-UHFFFAOYSA-N
Synonym 3,4-dichloro-6-nitroaniline, benzenamine, 4,5-dichloro-2-nitro, unii-3i0g2adu1l, 2-nitro-4,5-dichloroaniline, 3i0g2adu1l, 4,5-dichloro-2-nitro-aniline, 4,5-dichloro-2-nitrobenzenamine, 4,5-dichlor-2-nitroanilin, pubchem3466, acmc-1b6pu
PubChem CID 81149
IUPAC Name 4,5-dichloro-2-nitroaniline
SMILES C1=C(C(=CC(=C1Cl)Cl)[N+](=O)[O-])N

Specifications

Melting Point 180°C to 182°C
Quantity 5 g
Beilstein 643127
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material 4,5-Dichloro-2-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.