missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15415196
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1116681-96-9
C12H14BCl2FO2
290.95
MFCD16660310
PMLQYOJKCHQKNC-UHFFFAOYSA-N
2-4,5-dichloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,5-dichloro-2-fluorophenylboronic acid pinacol ester, 4,5-dichloro-2-fluorophenylboronic acid, pinacol ester, 4,5-dichloro-2-fluorophenylboronic acid,pinacol ester, 4,5-dichloro-2-fluorobenzeneboronic acid pinacol ester, 2-4,5-dichloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
53217250
2-(4,5-dichloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(Cl)C=C1F
This item is not returnable. View return policy
1116681-96-9
C12H14BCl2FO2
290.95
MFCD16660310
PMLQYOJKCHQKNC-UHFFFAOYSA-N
2-4,5-dichloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,5-dichloro-2-fluorophenylboronic acid pinacol ester, 4,5-dichloro-2-fluorophenylboronic acid, pinacol ester, 4,5-dichloro-2-fluorophenylboronic acid,pinacol ester, 4,5-dichloro-2-fluorobenzeneboronic acid pinacol ester, 2-4,5-dichloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
53217250
2-(4,5-dichloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(Cl)C=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 290.96
Percent Purity 96%
Chemical Name or Material 4,5-Dichloro-2-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'