missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™

Product Code. p-2701011
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10374222

Brand: Thermo Scientific Maybridge BTB07813DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 33098-21-4
Molecular Formula C11H8Cl2N2O
Molecular Weight (g/mol) 255.098
MDL Number MFCD00135290
InChI Key OWPCMQKRJFNLHF-UHFFFAOYSA-N
Synonym 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3-one, 4,5-dichloro-2-4-methylphenyl pyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-4-methylphenyl, 4,5-dichloro-2-p-tolyl-2h-pyridazin-3-one, 4,5-dichloro-2-4-methylphenyl-2-hydropyridazin-3-one, 4,5-dichloro-2-p-tolyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-3 2h-pyridazinone, 4,5-dichloro-2-4-methylphenyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3one
PubChem CID 2774757
IUPAC Name 4,5-dichloro-2-(4-methylphenyl)pyridazin-3-one
SMILES CC1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 255.1
Percent Purity 97%
Chemical Name or Material 4, 5-Dichloro-2-(4-methylphenyl)-2, 3-dihydropyridazin-3-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.