missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromothiophene-2-carboxylic acid, 98%, Thermo Scientific™

Product Code. 11439976
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11439976

Brand: Thermo Scientific Alfa Aesar B20928.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6324-10-3
MDL Number MFCD00039682

Specifications

Melting Point 228°C to 232°C
Odor Odorless
Molecular Formula C5H2Br2O2S
Quantity 25 g
Beilstein 135885
Synonym 4,5-dibromo-2-thiophenecarboxylic acid, 4,5-dibromothenoic acid, 4,5-dibromo-thiophene-2-carboxylic acid, pubchem7747, acmc-20a4op, maybridge1_002274, 4-dimethylaminoethyl benzoate, 4,5-dibromo-2-thiophencarboxylic acid, 2,3-dibromothiophene-5-carboxylic acid, 2-thiophenecarboxylicacid, 4,5-dibromo
InChI Key USFXRYVNRUMABJ-UHFFFAOYSA-N
SMILES C1=C(SC(=C1Br)Br)C(=O)O
IUPAC Name 4,5-dibromothiophene-2-carboxylic acid
Molecular Weight (g/mol) 285.94
PubChem CID 80611
Formula Weight 285.94
Percent Purity 98%
Chemical Name or Material 4,5-Dibromothiophene-2-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.