missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromo-2-furoic acid, 97%

Product Code. 11308306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308306 1 g 1g
11318306 5 g 5g
11328306 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11308306 Supplier Thermo Scientific Alfa Aesar Supplier No. L15517.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2434-03-9 | C5H2Br2O3 | 269.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2434-03-9
Molecular Formula C5H2Br2O3
Molecular Weight (g/mol) 269.88
MDL Number MFCD00092311
InChI Key BHUVICYZDBUMIU-UHFFFAOYSA-N
Synonym 4,5-dibromo-2-furoic acid, 2,3-dibromofuran-5-carboxylic acid, 4,5-dibromo-2-furancarboxylic acid, 2-furancarboxylicacid, 4,5-dibromo, 2-furancarboxylic acid, 4,5-dibromo, zlchem 604, acmc-209xd1, ksc201e3j, 4,5-dibromo-2-furoicacid
PubChem CID 4194832
IUPAC Name 4,5-dibromofuran-2-carboxylic acid
SMILES C1=C(OC(=C1Br)Br)C(=O)O

Specifications

Melting Point 169°C to 172°C
Quantity 1 g
Beilstein 129312
Formula Weight 269.88
Percent Purity 97%
Chemical Name or Material 4,5-Dibromo-2-furoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.