missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromo-2-fluorotoluene, 98%, Thermo Scientific™

Product Code. 15419968
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202982-77-2
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD00143234
InChI Key GJHJZTCWLKJMIG-UHFFFAOYSA-N
Synonym 4,5-dibromo-2-fluorotoluene, 2-fluoro-4,5-dibromotoluene, pubchem10411, benzene,1,2-dibromo-4-fluoro-5-methyl, 1,2-bis bromanyl-4-fluoranyl-5-methyl-benzene
PubChem CID 2773986
IUPAC Name 1,2-dibromo-4-fluoro-5-methylbenzene
SMILES CC1=CC(=C(C=C1F)Br)Br

Specifications

Quantity 1 g
Formula Weight 267.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4,5-Dibromo-2-fluorotoluene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'