missing translation for 'onlineSavingsMsg'
Learn More

4,5-Diazafluoren-9-one, 98%

Product Code. 15492448
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492448 1 g 1g
15402458 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492448 Supplier Thermo Scientific Alfa Aesar Supplier No. H56028.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50890-67-0 | C11H6N2O | 182.182 g/mol

4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].

Solubility
Soluble in dichloromethane, tetrahydrofuran, chloroform, benzene, toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.

Notes
Incompatible with strong oxidizing agents. Avoid heat and source of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50890-67-0
Molecular Formula C11H6N2O
Molecular Weight (g/mol) 182.182
MDL Number MFCD00046892
InChI Key PFMTUGNLBQSHQC-UHFFFAOYSA-N
Synonym 4,5-diazafluoren-9-one, 5h-cyclopenta 1,2-b:5,4-b' dipyridin-5-one, 5h-cyclopenta 2,1-b:3,4-b' dipyridin-5-one, 5h-pyrido 3',2':4,5 cyclopenta 1,2-b pyridin-5-one, pyridino 3',2'-1,2 cyclopenta 3,4-b pyridin-5-one, dafone, dafo, 4,5-diazafluorenone-0, 4,5-diaza-9h-fluoren-9-one
PubChem CID 342157
SMILES C1=CC2=C(C3=C(C2=O)C=CC=N3)N=C1

Specifications

Melting Point 214°C to 217°C
Quantity 1 g
Solubility Information Soluble in dichloromethane,tetrahydrofuran,chloroform,benzene,toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 4,5-Diazafluoren-9-one
compliance-icons
Product Identifier
  • 4,5-Diazafluoren-9-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4,5-Diazafluoren-9-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.