missing translation for 'onlineSavingsMsg'
Learn More

4-[[5-[[(Cyclopentyloxy)carbonyl]amino]-1-methyl-1H-indol-3-yl]methyl]-3-methoxybenzoic Acid, TRC

Product Code. 30136400
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136400 50 mg 50mg
30133413 100 mg 100mg
30005431 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136400 Supplier Toronto Research Chemicals Supplier No. TRCC98855050MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107754-20-1
Molecular Formula C24 H26 N2 O5
Molecular Weight (g/mol) 422.47
Synonym Benzoic acid, 4-[[5-[[(cyclopentyloxy)carbonyl]amino]-1-methyl-1H-indol-3-yl]methyl]-3-methoxy-, 4-[[5-[[(Cyclopentyloxy)carbonyl]amino]-1-methyl-1H-indol-3-yl]methyl]-3-methoxybenzoic acid
IUPAC Name 4-[[5-(cyclopentyloxycarbonylamino)-1-methylindol-3-yl]methyl]-3-methoxybenzoic acid
SMILES COc1cc(ccc1Cc2cn(C)c3ccc(NC(=O)OC4CCCC4)cc23)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H26N2O5/c1-26-14-17(11-15-7-8-16(23(27)28)12-22(15)30-2)20-13-18(9-10-21(20)26)25-24(29)31-19-5-3-4-6-19/h7-10,12-14,19H,3-6,11H2,1-2H3,(H,25,29)(H,27,28)
Formula Weight 422.1842
Chemical Name or Material 4-[(5-[((Cyclopentyloxy)carbonyl)amino]-1-methyl-1H-indol-3-yl)methyl]-3-methoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.