missing translation for 'onlineSavingsMsg'
Learn More

4',5,7-Trihydroxyisoflavone, 99+%

Product Code. 15485499
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485499 1 g 1g
15405509 100 mg 100mg
15495499 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15485499 Supplier Thermo Scientific Alfa Aesar Supplier No. J63241.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of tyrosine protein kinase | CAS: 446-72-0 | C15H10O5 | 270.24 g/mol

4',5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.

Solubility
Soluble in chloroform, methanol. Insoluble in cold water.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-72-0
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00016952
InChI Key TZBJGXHYKVUXJN-UHFFFAOYSA-N
Synonym genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
PubChem CID 5280961
ChEBI CHEBI:28088
IUPAC Name 5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

Melting Point ∼300°C
Color Brown to White
Quantity 1 g
Beilstein 263823
Merck Index 14,4391
Solubility Information Soluble in chloroform,methanol. Insoluble in cold water.
Formula Weight 270.24
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 4',5,7-Trihydroxyisoflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.