missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride, TRC

Product Code. 30413913
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413913 1 g -
30422950 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413913 Supplier Toronto Research Chemicals Supplier No. TRCT2957501G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28783-41-7
Molecular Formula C7 H9 N S . Cl H
Molecular Weight (g/mol) 175.68
Synonym Thieno[3,2-c]pyridine, 4,5,6,7-tetrahydro-, hydrochloride (1:1), Thieno[3,2-c]pyridine, 4,5,6,7-tetrahydro-, hydrochloride (8CI,9CI), 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
IUPAC Name 4,5,6,7-tetrahydrothieno[3,2-c]pyridine;hydrochloride
SMILES Cl.C1Cc2sccc2CN1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H9NS.ClH/c1-3-8-5-6-2-4-9-7(1)6;/h2,4,8H,1,3,5H2;1H
Formula Weight 175.0222
Chemical Name or Material 4,5,6,7-Tetrahydro-thieno[3,2-c]pyridine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.