missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride, Mikromol™

Product Code. 17990576
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17990576 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17990576 Supplier Mikromol Supplier No. MM0455.230025
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,5,6,7-Tetrahydro-thieno[3,2-c]pyridine Hydrochloride
CAS 28783-41-7
Analyte or Component Names 4,5,6,7-Tetrahydro-thieno[3,2-c]pyridine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C7 H9 N S . Cl H
Certifications/Compliance ISO 17025
Synonym Thieno[3,2-c]pyridine, 4,5,6,7-tetrahydro-, hydrochloride (1:1), Thieno[3,2-c]pyridine, 4,5,6,7-tetrahydro-, hydrochloride (8CI,9CI), 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
InChI Formula InChI=1S/C7H9NS.ClH/c1-3-8-5-6-2-4-9-7(1)6;/h2,4,8H,1,3,5H2;1H
SMILES Cl.C1Cc2sccc2CN1
IUPAC Name 4,5,6,7-tetrahydrothieno[3,2-c]pyridine;hydrochloride
Molecular Weight (g/mol) 175.68
Quantity 25 mg
Formula Weight 175.0222 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.