missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride, 96%, Alfa Aesar™

Product Code. 11357639
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357639 1 g 1g
11367639 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357639 Supplier Thermo Scientific Alfa Aesar Supplier No. H28243.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 28783-41-7
Molecular Formula C7H10ClNS
Molecular Weight (g/mol) 175.674
MDL Number MFCD05861485
InChI Key QMXCTKPNQFJZGK-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, 4,5,6,7-tetrahydrothieno 3,2,c pyridine hydrochloride, 4h,5h,6h,7h-thieno 3,2-c pyridine hydrochloride, 4,5,6,7-tetrahydrothieno 3,2-c pyridine hcl, 4,5,6,7-tetrahydrothieno 3,2,c pyridine hcl, unii-e0hb70dq7n, 4,5,6,7-tetrahydro-thieno 3,2-c pyridine hydrochloride, thieno 3,2-c pyridine, 4,5,6,7-tetrahydro-, hydrochloride
PubChem CID 10176342
IUPAC Name 4,5,6,7-tetrahydrothieno[3,2-c]pyridine;hydrochloride
SMILES C1CNCC2=C1SC=C2.Cl

Specifications

Melting Point 222°C to 224°C
Quantity 1 g
Formula Weight 175.68
Percent Purity 96%
Chemical Name or Material 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.