missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrothieno[3,2-c]pyridin-2-ol 2-Acetate Hydrochloride, TRC

Product Code. 30417307
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
5mg
Product Code. Quantity unitSize
30417307 50 mg -
30426223 5 mg 5mg
30409960 25 mg -
3 options
This item is not returnable. View return policy

Product Code. 30417307

Brand: Toronto Research Chemicals TRCT29581550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1151904-84-5
Formula Weight 233.0277
InChI Formula InChI=1S/C9H11NO2S.ClH/c1-6(11)12-9-4-7-5-10-3-2-8(7)13-9;/h4,10H,2-3,5H2,1H3;1H
IUPAC Name 4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate;hydrochloride
Molecular Formula C9H12ClNO2S
Molecular Weight (g/mol) 233.72
SMILES Cl.CC(=O)Oc1cc2CNCCc2s1
Synonym Thieno[3,2-c]pyridin-2-ol, 4,5,6,7-tetrahydro-, 2-acetate, hydrochloride (1:1) (ACI), 4,5,6,7-Tetrahydrothieno[3,2-c]pyridin-2-ol 2-acetate hydrochloride
Chemical Name or Material 4,5,6,7-Tetrahydrothieno[3,2-c]pyridin-2-ol 2-Acetate Hydrochloride

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.