missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 10332020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10332020 5 g 5g
10627040 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10332020 Supplier Thermo Scientific Acros Supplier No. 431090050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307308-03-8
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
InChI Key CSPINDKIWMCRNX-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carboxaldehyde, 4h,5h,6h,7h-pyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydro-pyrazolo 1,5-a pyridine-3-carbaldehyde, pyrazolo 1,5-a pyridine-3-carboxaldehyde,4,5,6,7-tetrahydro-9ci, pyrazolo 1,5-a pyridine-3-carboxaldehyde, 4,5,6,7-tetrahydro-9ci
PubChem CID 45083680
IUPAC Name 4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde
SMILES C1CCN2C(=C(C=N2)C=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 150.18
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.