missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene, 97%

Product Code. 15417257
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417257 250 mg 250mg
15497247 1 g 1g
15407257 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15417257 Supplier Thermo Scientific Alfa Aesar Supplier No. H34309.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13129-17-4 | C8H10S | 138.228 g/mol

It is an active pharmaceutical intermediate. Condensation of amino carboxyalkyl derivative of 4,5,6,7-tetrahydrobenzo[b]thiophene with salicylaldehyde yielded a potentially tridentate Schiff base (HSAT), which formed a series of copper(II) complexes. Reaction of ethyl 2-diazo-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 1 with 3-iminobutyronitrile 2 gave the hydrazone derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate. Condensation of amino carboxyalkyl derivative of 4,5,6,7-tetrahydrobenzo[b]thiophene with salicylaldehyde yielded a potentially tridentate Schiff base (HSAT), which formed a series of copper(II) complexes. Reaction of ethyl 2-diazo-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 1 with 3-iminobutyronitrile 2 gave the hydrazone derivative.

Notes
Air sensitive. Store at room temperature. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13129-17-4
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD19442759
InChI Key CBKDCOKSXCTDAA-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrobenzo b thiophene, benzothiophene, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-2-benzothiophene, benzo b thiophene, 4,5,6,7-tetrahydro, 5-17-01-00498 beilstein handbook reference
PubChem CID 22946
IUPAC Name 4,5,6,7-tetrahydro-1-benzothiophene
SMILES C1CCC2=C(C1)C=CS2

Specifications

Boiling Point 55°C (1 mmHg)
Refractive Index 1.558
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.