missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid, 97%

Product Code. 15448295
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448295 1 g 1g
15468295 250 mg 250mg
15458295 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448295 Supplier Thermo Scientific Alfa Aesar Supplier No. H33062.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40133-07-1 | C9H10O2S | 182.237 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40133-07-1
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00464469
InChI Key ROLXOQXKNDKXTA-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro, acmc-1asbb, enamine_001826, 2-carboxy-4,5,6,7-tetrahydro-1-benzothiophene, 2-carboxy-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylicacid, 4,5,6,7-tetrahydrobenzo b-thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic
PubChem CID 2063443
IUPAC Name 4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylic acid
SMILES C1CCC2=C(C1)C=C(S2)C(=O)O

Specifications

Melting Point 191°C to 193°C
Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 182.24
Percent Purity 97%
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.