missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydro-benzo[c]thiophene-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-3830088
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10752673 250 mg 250mg
10079131 1 g 1g
10352145 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10752673 Supplier Thermo Scientific Maybridge Supplier No. CC22801CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6435-75-2
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.24
MDL Number MFCD03086149
InChI Key ZBNTUDLPJCXPNF-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrobenzo c thiophene-1-carboxylic acid, 4,5,6,7-tetrahydro-benzo c thiophene-1-carboxylic acid, 1-carboxy-4,5,6,7-tetrahydrobenzo c thiophene, 4,5,6,7-tetrahydrobenzo 2,1-c thiophenecarboxylic acid, benzo c thiophene-1-carboxylicacid, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-2-benzo b thiophene-1-carboxylic acid
PubChem CID 299400
IUPAC Name 4,5,6,7-tetrahydro-2-benzothiophene-1-carboxylic acid
SMILES OC(=O)C1=C2CCCCC2=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 182.24
Percent Purity 97%
Chemical Name or Material 4,5,6,7-Tetrahydro-benzo[c]thiophene-1-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.