missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydro-5-(triphenylmethyl)thieno[3,2-c]pyridine, TRC

Product Code. 30423161
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423161 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423161 Supplier Toronto Research Chemicals Supplier No. TRCT29579550MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,5,6,7-Tetrahydro-5-(triphenylmethyl)thieno[3,2-c]pyridine
CAS 109904-25-8
Molecular Formula C26 H23 N S
Quantity 50 mg
Synonym Prasugrel Impurity 45
InChI Formula InChI=1S/C26H23NS/c1-4-10-22(11-5-1)26(23-12-6-2-7-13-23,24-14-8-3-9-15-24)27-18-16-25-21(20-27)17-19-28-25/h1-15,17,19H,16,18,20H2
SMILES C1Cc2sccc2CN1C(c3ccccc3)(c4ccccc4)c5ccccc5
IUPAC Name 5-trityl-6,7-dihydro-4H-thieno[3,2-c]pyridine
Molecular Weight (g/mol) 381.53
Formula Weight 381.1551
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.