missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydro-1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxo-1-piperidinyl)phenyl]-1H-Pyrazolo[3,4-c]pyridine-3-carboxylic Acid Methyl Ester, TRC

Product Code. 30420590
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420590 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420590 Supplier Toronto Research Chemicals Supplier No. TRCT29403010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,5,6,7-Tetrahydro-1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxo-1-piperidinyl)phenyl]-1H-Pyrazolo[3,4-c]pyridine-3-carboxylic Acid Methyl Ester
CAS 1074365-84-6
Molecular Formula C26 H26 N4 O5
Quantity 10 mg
Synonym 1H-Pyrazolo[3,4-c]pyridine-3-carboxylic acid','4,5,6,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxo-1-piperidinyl)phenyl]-','methyl ester (ACI), Methyl 4,5,6,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxo-1-piperidinyl)phenyl]-1H-pyrazolo[3,4-c]pyri
InChI Formula InChI=1S/C26H26N4O5/c1-34-20-12-10-19(11-13-20)30-24-21(23(27-30)26(33)35-2)14-16-29(25(24)32)18-8-6-17(7-9-18)28-15-4-3-5-22(28)31/h6-13H,3-5,14-16H2,1-2H3
SMILES COC(=O)c1nn(c2ccc(OC)cc2)c3C(=O)N(CCc13)c4ccc(cc4)N5CCCCC5=O
IUPAC Name methyl 1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate
Molecular Weight (g/mol) 474.508
Formula Weight 474.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.