missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Thiomorpholinyl)aniline, 97%

Product Code. 15450019
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450019 250 mg 250mg
15430019 1 g 1g
15440019 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450019 Supplier Thermo Scientific Alfa Aesar Supplier No. H63360.MD
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22589-35-1 | C10H14N2S | 194.296 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22589-35-1
Molecular Formula C10H14N2S
Molecular Weight (g/mol) 194.296
MDL Number MFCD01625958
InChI Key RQXQMUUSQGCLPL-UHFFFAOYSA-N
Synonym 4-thiomorpholinoaniline, 4-thiomorpholin-4-yl aniline, 4-thiomorpholin-4-yl phenylamine, 4-thiomorpholinobenzenamine, 4-thiomorpholin-4-yl-phenylamine, benzenamine,4-4-thiomorpholinyl, benzenamine, 4-4-thiomorpholinyl, 4-p-aminophenyl-tetrahydro-4h-1,4-thiazine
PubChem CID 10774206
IUPAC Name 4-thiomorpholin-4-ylaniline
SMILES C1CSCCN1C2=CC=C(C=C2)N

Specifications

Quantity 250 mg
UN Number UN2811
Formula Weight 194.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Thiomorpholinyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.