missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Thiodiphenol, 99%, Thermo Scientific™

Product Code. 11955731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11955731 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11955731 Supplier Thermo Scientific Acros Supplier No. 201970250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2664-63-3
CAS Min % 98.5
CAS Max % 100.0
Melting Point 151.0°C to 154.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C12H10O2S
Linear Formula S(C6H4OH)2
MDL Number MFCD00002349
Quantity 25 g
Beilstein 06,86
Synonym 4,4'-thiodiphenol, bis 4-hydroxyphenyl sulfide, thiobisphenol, 4,4'-thiobis-phenol, phenol, 4,4'-thiobis, 4,4'-thiobisphenol, 4,4'-dihydroxydiphenyl sulfide, bis 4-oxyphenyl sulfide, p,p'-dihydroxydiphenyl sulfide, 4,4'-dioxydiphenyl sulfide
InChI Key VWGKEVWFBOUAND-UHFFFAOYSA-N
SMILES OC1=CC=C(SC2=CC=C(O)C=C2)C=C1
IUPAC Name 4-(4-hydroxyphenyl)sulfanylphenol
Molecular Weight (g/mol) 218.27
PubChem CID 17570
ChEBI CHEBI:38957
Formula Weight 218.28
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Thiodiphenol
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Thiodiphenol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.