missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(tert-Butyl)-1,3-thiazol-2-yl]benzaldehyde, 97%, Thermo Scientific™

Product Code. p-2701163
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10663593 250 mg 250mg
10099251 1 g 1g
10409084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10663593 Supplier Thermo Scientific Maybridge Supplier No. KM04244CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-78-5
Molecular Formula C14H15NOS
Molecular Weight (g/mol) 245.34
MDL Number MFCD00084914
InChI Key IIKIVYFYAIKVBC-UHFFFAOYSA-N
Synonym 4-4-tert-butyl-1,3-thiazol-2-yl benzaldehyde, benzaldehyde,4-4-1,1-dimethylethyl-2-thiazolyl, 4-tert-butyl-2-4-formylphenyl thiazole, 4-4-tert-butyl-2-thiazolyl benzaldehyde, 4-4-tert-butylthiazol-2-yl benzaldehyde, 4-4-tert-butyl thiazol-2-yl benzaldehyde
PubChem CID 2778859
IUPAC Name 4-(4-tert-butyl-1,3-thiazol-2-yl)benzaldehyde
SMILES CC(C)(C)C1=CSC(=N1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 245.34
Percent Purity 97%
Chemical Name or Material 4-[4-(tert-Butyl)-1, 3-thiazol-2-yl]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.