missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(tert-Butoxycarbonyl)piperazino]benzoic acid, 97%, Thermo Scientific™

Catalog Number 10012212
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
162046-66-4
C16H22N2O4
306.362
BEDWYXZFIYMEJG-UHFFFAOYSA-N
4-4-tert-butoxycarbonyl piperazin-1-yl benzoic acid, 4-4-tert-butoxycarbonyl piperazino benzoic acid, 4-4-carboxyphenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-1-piperazineyl benzoic acid, 4-4-carboxy-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-4-carboxyphenyl-4-boc piperazine, 4-4-boc-piperazin-1-yl benzoic acid, 1-boc-4-4-carboxy-phenyl-piperazine, 1-4-boc-piperazin-1-yl-benzoic acid
2795508
4-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]benzoic acid
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C(=O)O
This item is not returnable. View return policy
162046-66-4
C16H22N2O4
306.362
BEDWYXZFIYMEJG-UHFFFAOYSA-N
4-4-tert-butoxycarbonyl piperazin-1-yl benzoic acid, 4-4-tert-butoxycarbonyl piperazino benzoic acid, 4-4-carboxyphenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-1-piperazineyl benzoic acid, 4-4-carboxy-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-4-carboxyphenyl-4-boc piperazine, 4-4-boc-piperazin-1-yl benzoic acid, 1-boc-4-4-carboxy-phenyl-piperazine, 1-4-boc-piperazin-1-yl-benzoic acid
2795508
4-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]benzoic acid
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C(=O)O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 306.36
Percent Purity 97%
Chemical Name or Material 4-[4-(tert-Butoxycarbonyl)piperazino]benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'