missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Sulfonyldiphenol, 99.7%

Product Code. 10466970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10466970 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10466970 Supplier Thermo Scientific Acros Supplier No. 146915000
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-09-1 | C12H10O4S | 250.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 80-09-1
CAS Min % 99.6
CAS Max % 100.0
Melting Point >245°C
Color Gray-Green to White
Infrared Spectrum Authentic
Assay Percent Range 99.6 % min. (HPLC)
Packaging Plastic bottle
Molecular Formula C12H10O4S
Linear Formula O2S(C6H4OH)2
MDL Number MFCD00002350
Quantity 500 g
Beilstein 06, 861
Synonym 4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c
Solubility Information Solubility in water: 1.1g/L (20°C). Other solubilities: soluble in alcohol and acetone
InChI Key VPWNQTHUCYMVMZ-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1
IUPAC Name 4-(4-hydroxyphenyl)sulfonylphenol
Molecular Weight (g/mol) 250.27
PubChem CID 6626
ChEBI CHEBI:34372
Formula Weight 250.27
Percent Purity 99.70%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Sulfonyldiphenol, 99.7%
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Sulfonyldiphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.