missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridylmethyl)aniline, 97%, Thermo Scientific™

Product Code. 10059191
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10059191 1 g Amber Glass Bottle 1g
10344702 250 mg Amber Glass Bottle 250mg
10615145 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10059191 Supplier Thermo Scientific Maybridge Supplier No. BTB01214DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 27692-74-6
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00053060
InChI Key WZXYYQHVDJMIFF-UHFFFAOYSA-N
Synonym 4-4-pyridylmethyl aniline, 4-pyridin-4-ylmethyl aniline, 4-pyridin-4-ylmethyl-phenylamine, benzenamine, 4-4-pyridinylmethyl, 4-4-pyridinylmethyl aniline, 4-pyridin-4-yl methyl aniline, 4-pyridin-4-ylmethyl phenyl amine, 4-4-pyridylmethyl phenylamine, 4-p-aminobenzylpyridine, enamine_005904
PubChem CID 307158
IUPAC Name 4-(pyridin-4-ylmethyl)aniline
SMILES C1=CC(=CC=C1CC2=CC=NC=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.24
Percent Purity 97%
Grade Technical
Chemical Name or Material 4-(4-Pyridylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.