missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Oxydibenzenesulfonyl Hydrazide, TRC

Product Code. 30423130
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423130 1 g 1g
30422998 5 g -
30416806 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423130 Supplier Toronto Research Chemicals Supplier No. TRCO8706381G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-51-3
Molecular Formula C12H14N4O5S2
Molecular Weight (g/mol) 358.39
Synonym 4,4'-Oxybis(benzenesulfohydrazide), 4,4'-Oxybis(benzenesulfonic Acid Hydrazide), 4,4'-Oxybis(benzenesulfonic Acid) Dihydrazide, 4,4'-Oxybis(benzenesulfonyl hydrazide), 4,4'-Oxydi(benzenesulfonic Acid Hydrazide), 4,4'-Oxydibenzenesulfonic Acid Dihydrazide, 4,4'-Oxydibenzenesulfonyl Hydrazide, Neocellborn P 100, Neocellborn P 1000, Neocellborn P 1000M, Neothlene HM 806P, Neothlene HM 80NP, Unifoam AZ 90, p,p'-Oxybis(benzenesulfohydrazide), p,p'-Oxybis(benzenesulfone hydrazide), p,p'-Oxybis(benzenesulfonyl hydrazide), p,p'-Oxybis(benzenesulfonylhydrazine), S 250 (blowing agent), Tracel OBSH, Tracel OBSH 160NER, Tracel OBSH 80PR
IUPAC Name 4-[4-(hydrazinesulfonyl)phenoxy]benzenesulfonohydrazide
SMILES NNS(=O)(=O)c1ccc(Oc2ccc(cc2)S(=O)(=O)NN)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H14N4O5S2/c13-15-22(17,18)11-5-1-9(2-6-11)21-10-3-7-12(8-4-10)23(19,20)16-14/h1-8,15-16H,13-14H2
Formula Weight 358.0406
Chemical Name or Material 4,4'-Oxydibenzenesulfonyl Hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.