missing translation for 'onlineSavingsMsg'
Learn More

4,4’-Oxydibenzenesulfonyl Chloride, TRC

Product Code. 30421592
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421592 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421592 Supplier Toronto Research Chemicals Supplier No. TRCO8706305G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Oxydibenzenesulfonyl Chloride
CAS 121-63-1
Molecular Formula C12H8Cl2O5S2
Quantity 5 g
Synonym 4,4'-Oxybis[benzenesulfonyl Chloride], 4,4'-Bis(chlorosulfonyl)biphenyl Ether, 4,4'-Bis(chlorosulfonyl)diphenyl Ether, 4,4'-Oxybis(benzenesulfonyl Chloride), 4,4'-Oxydibenzenesulfonyl Chloride, Bis(4-Chlorosulfonylphenyl) Ether, Diphenyl Ether 4,4’-Disulfonyl Chloride, NSC 212, Oxybis(4-benzenesulfonyl Chloride), Oxybis(benzenesulfonyl Chloride), Phenoxybenzene-4,4'-disulfonyl Chloride, p,p'-Oxybis(benzenesulfonyl Chloride), 4,4’-Oxybisbenzenesulfonyl Chloride
InChI Formula InChI=1S/C12H8Cl2O5S2/c13-20(15,16)11-5-1-9(2-6-11)19-10-3-7-12(8-4-10)21(14,17)18/h1-8H
SMILES ClS(=O)(=O)c1ccc(Oc2ccc(cc2)S(=O)(=O)Cl)cc1
IUPAC Name 4-(4-chlorosulfonylphenoxy)benzenesulfonyl chloride
Molecular Weight (g/mol) 367.22
Formula Weight 365.919
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.