missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Oxybis(benzoic acid), 98+%, Thermo Scientific Chemicals

Product Code. 11372727
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2215-89-6
Molecular Formula C14H10O5
Molecular Weight (g/mol) 258.229
MDL Number MFCD00013988
InChI Key WVDRSXGPQWNUBN-UHFFFAOYSA-N
Synonym 4,4'-oxybis benzoic acid, 4,4'-oxydibenzoic acid, 4,4'-oxybisbenzoic acid, 4-4-carboxyphenoxy benzoic acid, benzoic acid, 4,4'-oxybis, 4,4'-dicarboxydiphenyl ether, 4,4-oxobisbenzoic acid, 4,4'-diphenyl ether dicarboxylic acid, diphenyl ether 4,4'-dicarboxylic acid, pubchem9032
PubChem CID 75183
IUPAC Name 4-(4-carboxyphenoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 329°C to 332°C
Quantity 25 g
Beilstein 2218315
Formula Weight 258.23
Percent Purity ≥98%
Chemical Name or Material 4,4'-Oxybis(benzoic acid)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'